missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Economical pH Test Strips
Line of pH test strips incorporates the most up-to-date technology.
Supplier: Customer Channels Group (Ccg) PH00151B50
Description
- Nonbleed paper padded on a vinyl, 2.5 in. (6.35cm) wand
- Stabilized to provide the most accurate color determinations
- Color charts printed on each pack for quick and easy identification
- Each pack includes 50 strips
Chemical Identifiers
| Stabilized to provide user with most accurate color determination | |
| 50 Bag | |
| pH Test Strip |
Specifications
| 0 to 1.5 | |
| 0.5 | |
| pH Test Strip |
| Stabilized to provide user with most accurate color determination | |
| 50 Bag |
Safety and Handling
ShelfLife : 3 Years