missing translation for 'onlineSavingsMsg'
Learn More

Econazole nitrate

Catalog No. p-7076606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6317306 5 g
AAJ6317314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6317306 Supplier Thermo Scientific Chemicals Supplier No. J6317306

CAS: 24169-02-6 | C18H16Cl3N3O4 | 444.693 g/mol

Econazole nitrate is an antifungal medication often used as a cream to treat skin infections like tinea, ringworm and athlete's foot, personal care, cosmetic and pharmaceutical applications. It is used for studies on processes such as platelet serotonin uptake, prostanoid biosynthesis, EDHF-mediated relaxation, and Ca2+ transport pathways in thymic lymphocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24169-02-6
Molecular Formula C18H16Cl3N3O4
Molecular Weight (g/mol) 444.693
MDL Number MFCD00058160
InChI Key DDXORDQKGIZAME-UHFFFAOYSA-N
Synonym econazole nitrate, ifenec, epi-pevaryl, gyno-pevaryl, econazole nitrate salt, pevaryl, +--econazole nitrate, spectazole, econazolum nitrate, gyno-pevaryl 150
PubChem CID 68589
IUPAC Name 1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole;nitric acid
SMILES C1=CC(=CC=C1COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl)Cl.[N+](=O)(O)[O-]

Specifications

Melting Point 158°C to 163°C
Packaging Plastic bottle
Quantity 5 g
Solubility Information Soluble at 0.4mg/ml in DMSO. Sparingly soluble in water.
Formula Weight 444.7
Physical Form Powder
Chemical Name or Material Econazole nitrate
Hazard Category H302-H315-H319
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 246-053-5
RTECSNumber NI4450000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only