Learn More
Berkshire™ EcoClean™ 60
Berkshireâs EcoClean™ 60 line of products is a new class of nonwoven materials that is 100% biodegradable and sustainable 'green' product. The EcoClean™ wiper is composed of U.S.-sourced cotton fibers and is manufactured by a proprietary process for low fiber release and high strength.
Supplier: Berkshire™ ECW60121220
Description
- Excellent strength
- Heat resistant
- Highly absorbent and durable
- Extremely soft
- Resists build up of static charge
Specifications
| Surface Wiper | |
| 30 cm | |
| Non-woven Cotton | |
| ISO Class 5-8 |
| 12 in. | |
| 30 cm | |
| White |