Learn More
LevGo Eco-smartFunnel™ Disposable Eco Friendly Paper Lab Funnel
Eco-smartFunnel is a simple, cost-effective solution to reduce plastic use in the lab and reduce cross contamination from reuse of washed funnels
Supplier:  LevGo 19620
 View more versions of this product
Catalog No. 15555002
Only null left
Description
Description
  • Reduce cross contamination
  • Fold flat design uses less storage space
  • Great for powders and cold liquids
  • Can be used in Saliva Collection Applications
  • Eco friendly, biodegradable
  • Made in the USA
Specifications
Specifications
Paper Disposable Funnel
FSC Certified Food grade clay coated paperboard
Square
2.75 x 1.12 in.
Single-use
Sample collection, pouring viscous liquids and powders
Funnel
1.6 g
Product Suggestions
Product Suggestions