Learn More
BTX™ ECM™ 830 Square Post Adapters to Banana Plug Cables (45-0217)
Catalog No. BTX450089
Change view
Click to view available options
Connection Type:
Banana Cables
Catalog No. Connection Type
BTX450089 Banana Cables
1 options
Catalog No. BTX450089 Supplier BTX™ Supplier No. 450089
ECM™ 830 Square Post Adapters to Banana Plug Cables (45-0217) for use with ECM™ 830.
  • For connection to electroporation generators
  • Square Post Adapters
Specifications
For Use With (Application) To connect an electrofusion/electroporation system to the electrodes and chambers to be used for a particular application
For Use With (Equipment) Banana Plug Cables 45-0217
Product Type Square Post Adapters
Description Square Post Adapters, Red and Black, to Banana Plug Cables 45-0217
Display LCD
Sterility Non-sterile
Connection Type Banana Cables
Interface Digital User
Pulse Form Constant, Linear, Non-linear