missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECL Plex Goat anti-Rabbit IgG-Cy™ 5 Secondary Antibody, Cytiva
$254.00
Specifications
| Antigen | IgG |
|---|---|
| Classification | Monoclonal |
| Conjugate | Cyanine5 |
| Form | Purified |
| Host Species | Goat |
Description
Host Species: Goat
Isotype: IgG
Species Reactivity: Rb
Western Blotting
Specifications
| IgG | |
| Cyanine5 | |
| Goat | |
| Secondary |
| Monoclonal | |
| Purified | |
| IgG |