missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Echinacoside, ≥98% (HPLC), MilliporeSigma™ Supelco™
Description
Echinacoside (ECH) is a typical phenylethanoid glycoside. It is known to contain multiple unstable chemical bonds and high reactive functional groups which aids to pathways involving degradation and metabolism. It is isolated from the stems of Cistanches salsa. It is a crude drug mostly used as antisenium and antifatigue agents. Featured Industries: Food and Beverages
Specifications
Specifications
| CAS | 82854-37-3 |
| Linear Formula | C35H46O20 |
| MDL Number | MFCD00075695 |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 786.73 |
| Percent Purity | ≥98% (HPLC) |
| Grade | Analytical Standard |