missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecamsule, ≥97.0% (T); ≥98.0% (HPLC), MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 92761-26-7 |
| Physical Form | Neat |
| Linear Formula | C28H34O8S2 |
| Synonym | 3,3 ′-(1,4-Phenylenedimethylidyne)bis[7,7-dimethyl-2-oxo-bicyclo[2.2.1 ]heptane-1-methanesulfonic acid]; Terephthalylidene-3,3 ′-dicamphor-10,10 ′-disulfonic acid |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 562.69 |
| Percent Purity | ≥97.0% (T); ≥98.0% (HPLC) |
| Grade | Analytical Standard |