missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin Super Bright 436 Conjugate

Catalog No. 62431782
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62431782 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 62431782 Supplier Invitrogen™ Supplier No. 62431782
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

eBioscience Streptavidin Super Bright 436 is a streptavidin-fluorochrome conjugate for indirect staining protocols to detect biotinylated primary antibodies in flow cytometry.

Super Bright dyes are polymer-based fluorophores that are excited by the violet laser (405 nm) and emit fluorescence at the wavelength indicated in their name. They are optimized for use in flow cytometry and allow for use of additional channels with violet excitation. Super Bright dyes are fully compatible with other commonly used fluorescent molecules, eBioscience buffers and fixatives, viability stains, and reagents.

Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.

Applications tested
Streptavidin Super Bright 436 Conjugate has been tested by flow cytometric analysis to detect biotinylated primary antibodies. The amount of this conjugate that will stain a cell sample in a final volume of 100 μL is less than or equal to 0.125 μg. Cell number should be determined empirically but can range from 105 to 108 cells/test. It is recommended that the streptavidin be carefully titrated for optimal performance in the assay of interest.

When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Complete Staining Buffer for more information.

Super Bright polymer dyes are sold under license from Becton, Dickinson and Company.

Specifications

Concentration 0.2 mg/mL
Conjugate Super Bright 436
Quantity 100 μg
Content And Storage Store in refrigerator (2°C to 8°C).

Shipping Condition Wet Ice
Form Liquid
Product Line eBioscience
Product Type Streptavidin Conjugate (Fluorescent)
Target Molecule Biotin
Label or Dye Super Bright 436

For Research Use Only. Not for use in diagnostic procedures.