missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin PE-eFluor™ 610 Conjugate

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.

$492.00 - $492.00

Specifications

Concentration 0.2 mg/mL
Conjugate PE-eFluor 610
Form Liquid
Product Line eBioscience™
Product Type Streptavidin Conjugate (Fluorescent)
View More Specs

Alert:

Contains Sodium azide


Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124814
View Documents
Invitrogen™
61431782
100 μg N/A
Only null left
 
Description

Description

Specifications

Specifications

0.2 mg/mL
Liquid
Streptavidin Conjugate (Fluorescent)
PE-eFluor 610
PE-eFluor 610
eBioscience™
Biotin
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'