missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Phalloidin eFluor™ 570

Catalog No. 501123637
Change view
Click to view available options
Quantity:
300 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501123637 300 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501123637 Supplier Invitrogen™ Supplier No. 41655905

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phalloidin is a peptide toxin isolated from the Amanita phalloides mushroom. Phalloidin selectively binds F-actin and functions to stablilize actin polymers by inhibiting the dissociation of actin monomers from the filament ends.

Phalloidin is a peptide toxin isolated from the Amanita phalloides mushroom. Phalloidin selectively binds F-actin and functions to stablilize actin polymers by inhibiting the dissociation of actin monomers from the filament ends. Because F-actin polymers compose the cytoskeleton of almost all eukaryotic cells, phalloidin conjugates can be useful probes for visualizing the structure of live or fixed cells using microscopy. Phalloidin eFluor™ 570 can be used to label cells of multiple species.

Phalloidin eFluor™ 570 is supplied as a frozen liquid. To prepare the 1000X stock solution, thaw the vial, add 30 μL high-quality anhydrous DMSO, mix and briefly spin down. The resulting solution (phalloidin DMSO stock) should be aliquoted, protected from light and moisture, and stored at -20°C. To prepare a 1X working solution of phalloidin conjugate, add 1 μL of 1000X phalloidin DMSO stock solution to 1 mL of PBS containing 1% BSA.

Reported Applications

  • Immunocytochemistry
  • Immunohistochemical Staining
  • Microscopy

Specifications

For Use With (Application) Immunocytochemistry, Immunohistochemical Staining, Microscopy
Quantity 300 tests
Excitation Wavelength Range 555/570 nm
Dye Type eFluor 570
Concentration 1000X
Source A peptide toxin isolated from the Amanita phalloides mushroom
No. of Tests 300 Tests
Content And Storage ≤-20°C. Protect from light and moisture.
Product Type Phalloidin
Form Frozen Liquid
Product Line eBioscience
Label Type eFluor
Sub Cellular Localization Cytoskeleton
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.