missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Essential Human Th1/Th17 Phenotyping Kit DFS Item

Invitrogen™ eBioscience™ Essential Human Th1/Th17 Phenotyping Kit is designed for flow cytometry-based identity testing of Type 1 and Type 17 T-helper cells

Supplier:  Invitrogen™ A42927

Catalog No. A42927


Only null left
Explore available promotions

Description

Description

Phenotyping Kit leverages high quality eBioscience™ antibodies, isotype controls, cell stimulation reagents (induces cytokine production), and fixation/permeabilization reagents (for intracellular markers IfN-g and IL-17) and includes a ready-to-use protocol for experimental setup, gating strategy, and analysis.

Benefits include:

  • Proven performance―antibodies and supporting reagents designed for flow cytometry
  • Highly referenced antibodies―includes one of the most referenced clones for IL-17
  • Turnkey kit and protocol―contains reagents and standardized gating recommendations needed to perform flow cytometry characterization
Specifications

Specifications

  • CD4, Ifn-g, IL-17a antibodies, store at 2°C to 8°CC
  • Cell stimulating cocktail, store at -5 to -30°C
  • Protein transport cocktail, store at -5 to -30°C
  • Intracellular fixation & permealization reagents, store at 2°C to 8°CC
  • Isotype control, store at 2°C to 8°CC
100 reactions
Approved for shipment at Room Temperature or on Wet or Dry Ice
Phenotyping Kit
Kit
Fluorescence
eBioscience
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.