missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Concanavalin A (Con A) Solution (500X)
Description
Con A is isolated from the Jack bean, Canavlia ensiformis. It is a lectin that binds alpha-D-glucose and alpha-D-mannose moieties found in various glycoproteins, glycolipids, and sugars and is a potent leukocyte mitogen. This reagent is intended for use in in vitro activation of human and mouse leukocytes.
Reported Application
Functional Assays
Specifications
Specifications
| Format | Vial |
| Quantity | 4 x 100 μL |
| Source | Canavlia Ensiformis |
| Form | Solution |
| Protein Subtype | Lectins |
| Product Line | eBioscience |
For Research Use Only. Not for use in diagnostic procedures.