missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Avidin HRP
Description
Avidin horseradish peroxidase is commonly used as a second step for the detection of biotinylated antibodies. Avidin is a glycoprotein from chicken egg whites that binds biotin with high affinity. Biotin is a tag that is frequently used to label antibodies and other probes for immunodetection methods.
This product is recommended for use in immunohistochemistry applications.
Conjugate: HRP
Reported Application: Immunohistochemical Staining
Specifications
Specifications
| Target Molecule | Biotin |
| Quantity | 5 x 100 μL |
| Content And Storage | Store at 2°C to 8°C |
| Formulation | Aqueous buffer, contains carrier protein/stabilizer, no sodium azide |
| For Use With (Application) | Immunohistochemical Staining |
| Product Line | eBioscience |
| Label or Dye | HRP (Horseradish Peroxidase) |
For Research Use Only. Not for use in diagnostic procedures.