missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Avidin HRP

Catalog No. 501123248
Change view
Click to view available options
Quantity:
100 μL
5 x 100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501123248 5 x 100 μL
501123249 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 501123248 Supplier Invitrogen™ Supplier No. 18420089

Avidin horseradish peroxidase is commonly used as a second step for the detection of biotinylated antibodies.

Avidin horseradish peroxidase is commonly used as a second step for the detection of biotinylated antibodies. Avidin is a glycoprotein from chicken egg whites that binds biotin with high affinity. Biotin is a tag that is frequently used to label antibodies and other probes for immunodetection methods.

This product is recommended for use in immunohistochemistry applications.

Conjugate: HRP

Reported Application: Immunohistochemical Staining

Specifications

Target Molecule Biotin
Quantity 5 x 100 μL
Content And Storage Store at 2°C to 8°C
Formulation Aqueous buffer, contains carrier protein/stabilizer, no sodium azide
For Use With (Application) Immunohistochemical Staining
Product Line eBioscience
Label or Dye HRP (Horseradish Peroxidase)

For Research Use Only. Not for use in diagnostic procedures.