missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Ebastine | |
| 87°C | |
| C32H39NO2 | |
| 1 g | |
| MJJALKDDGIKVBE-UHFFFAOYSA-N | |
| 4-(4-benzhydryloxypiperidin-1-yl)-1-(4-tert-butylphenyl)butan-1-one | |
| 3191 | |
| ≥98.0% (HPLC,T) |
| 90729-43-4 | |
| White | |
| MFCD00865661 | |
| ebastine, kestine, ebastel, ebastin, kestin, evastel, bactil, ebastinum latin, las w-090, ebastina spanish | |
| CC(C)(C)C1=CC=C(C=C1)C(=O)CCCN2CCC(CC2)OC(C3=CC=CC=C3)C4=CC=CC=C4 | |
| 469.669 | |
| 469.67 | |
| Crystalline Powder |
Chemical Identifiers
| 90729-43-4 | |
| 469.669 | |
| MJJALKDDGIKVBE-UHFFFAOYSA-N | |
| 3191 | |
| CC(C)(C)C1=CC=C(C=C1)C(=O)CCCN2CCC(CC2)OC(C3=CC=CC=C3)C4=CC=CC=C4 |
| C32H39NO2 | |
| MFCD00865661 | |
| ebastine, kestine, ebastel, ebastin, kestin, evastel, bactil, ebastinum latin, las w-090, ebastina spanish | |
| 4-(4-benzhydryloxypiperidin-1-yl)-1-(4-tert-butylphenyl)butan-1-one |
Safety and Handling
RTECSNumber : EL8140000
TSCA : No