Learn More
Interscience EasySpiral
Catalog No. 16949051
Change view
Click to view available options
Product Line:
EasySpiral
EasySpiral Dilute
EasySpiral Pro
EasySpiral Pro Milk
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Line
16949051 EasySpiral Pro
16949030 EasySpiral
16948919 EasySpiral Pro Milk
16948888 EasySpiral Dilute
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 16949051 Supplier Interscience Supplier No. 413000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The easySpiral plater uses the Spiral technique for plating Petri dishes.
EasySpiral is an automatic plater of Petri dishes. It allows plating of a sample with 300 to 1.3x10^5 CFU/mL on one Petri dish, without prior dilution. Plated volumes are of 50 and 100 μL.
  • Syringe capacity: 1000 μL
  • Plating modes: exponential, circle
  • Counting range: from 300 to 1.3x10^5 CFU/mL
  • Plating cycle time: 25 seconds
EasySpiral Pro is an automatic plater of Petri dishes. It allows plating of a sample of 100 to 1x10^7 CFU/mL on one Petri dish, without prior dilution. Traceability is ensured and volumes are programmable via USB connection.
  • Traceability and programmable volumes
  • Plating modes: exponential, constant, circle
  • Counting range: from 100 to 1x10^7 CFU/mL
  • Plating cycle time: 25 seconds
EasySpiral Pro Milk is an automatic Petri dish plater. It allows plating of a sample of 100 to 1x10^7 CFU/mL on a single Petri dish, without prior dilution. EasySpiral Pro Milk is particularly suitable for the dairy industry in analyzing raw materials and payment for milk.
  • Sample intake in the sample holder
  • Plating modes: exponential, pour, circle
  • Counting range: from 100 to 1x10^7 CFU mL
  • Plating cycle time: 25 seconds
EasySpiral Dilute is an automatic diluter and plater of Petri dishes. It allows 1/10th serial dilutions and plating up to 6 log. dilution on 1 single Petri dish. For a sample of 30 to 1 x 10^12 bacteria, no more manual dilutions and plating is automatic.
  • Automatic cycle of 5 successive dilutions at 1/10th
  • Plating modes: exponential, constant, circle, pour plate
  • Count range: from 30 to 1x10^12 CFU/mL
  • Plating cycle time: 134 seconds
Specifications
Type Automatic Plater
Product Line EasySpiral Pro
Speed 1 Cycle/25 sec.
Voltage 110/240 V