Learn More
Interscience EasySpiral
The easySpiral plater uses the Spiral technique for plating Petri dishes.
Supplier: Interscience 413090
Description
EasySpiral is an automatic plater of Petri dishes. It allows plating of a sample with 300 to 1.3x10^5 CFU/mL on one Petri dish, without prior dilution. Plated volumes are of 50 and 100 μL.
- Syringe capacity: 1000 μL
- Plating modes: exponential, circle
- Counting range: from 300 to 1.3x10^5 CFU/mL
- Plating cycle time: 25 seconds
EasySpiral Pro is an automatic plater of Petri dishes. It allows plating of a sample of 100 to 1x10^7 CFU/mL on one Petri dish, without prior dilution. Traceability is ensured and volumes are programmable via USB connection.
- Traceability and programmable volumes
- Plating modes: exponential, constant, circle
- Counting range: from 100 to 1x10^7 CFU/mL
- Plating cycle time: 25 seconds
EasySpiral Pro Milk is an automatic Petri dish plater. It allows plating of a sample of 100 to 1x10^7 CFU/mL on a single Petri dish, without prior dilution. EasySpiral Pro Milk is particularly suitable for the dairy industry in analyzing raw materials and payment for milk.
- Sample intake in the sample holder
- Plating modes: exponential, pour, circle
- Counting range: from 100 to 1x10^7 CFU mL
- Plating cycle time: 25 seconds
EasySpiral Dilute is an automatic diluter and plater of Petri dishes. It allows 1/10th serial dilutions and plating up to 6 log. dilution on 1 single Petri dish. For a sample of 30 to 1 x 10^12 bacteria, no more manual dilutions and plating is automatic.
- Automatic cycle of 5 successive dilutions at 1/10th
- Plating modes: exponential, constant, circle, pour plate
- Count range: from 30 to 1x10^12 CFU/mL
- Plating cycle time: 134 seconds
Chemical Identifiers
| Automatic Plater |
Specifications
| Automatic Plater | |
| EasySpiral Pro Milk | |
| 1 Cycle/25 sec. | |
| 100/240 V |