Learn More
Mettler Toledo™ EasyDirect Moisture
Catalog No. 01912576
Change view
Click to view available options
For: HS/HX Moisture Anlayzers
Catalog No.
01912576 For: HS/HX Moisture Anlayzers
1 options
Catalog No. 01912576 Supplier Mettler Toledo™ Supplier No. 30522676
Simple Data Management Software. Data collection in SQL database, graphical result evaluation, central instrument management with routine testing scheduler.
  • Secure Data Management
  • Connect up to 5 Moisture Analyzers
  • Routine Testing Scheduler
Specifications
For Use With (Equipment) HS/HX Moisture Analyzer
Type Moisture Analyzer Software