missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Easy-Adapt™ CHO K1 CD Medium
GREENER_CHOICE

Catalog No. 670114
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
670114 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 670114 Supplier Gibco™ Supplier No. 670114

Chemically defined (CD) medium

Rich in carbohydrates, amino acids, and vitamins, and specifically designed to provide the nutrients CHO cells need.

Specifications

Content And Storage Storage conditions: 2°C to 8°C. Protect from light.
Cell Line CHO, CHO-K1
Classification Animal Origin-free, Chemically-defined, Protein-free
Form Powder
Product Type Cell Culture Media
Format BPC
Quantity 20 L
Source Animal origin-free