missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Earthworm, Cross Section

Catalog No. S30346
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30346 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30346 Supplier Eisco™ Supplier No. BS18198

Prepared microscope slide of a cross section of earthworm. Stained for better visualization of characteristic structures. Ideal for biology classrooms to explore structure-function relationships as per NGSS standards.

  • Pre-cleaned glass slide with ground edges
  • Size: 1 x 3 in. (25 x 77 mm)
  • Professionally stained samples for best visualization
  • Ideal for biology classrooms to explore structure-function relationships as per NGSS standards
  • Shows characteristic structures

Specifications

Biosafety Level 1
Concentration 10%
Classification General
Material Glass
Type Prepared Microscope Slide
Quantity 1 Ea.
Slide Type Prepared Slide