Learn More
Invitrogen™ Ea52-68 peptide bound to I-Ab Monoclonal Antibody (eBioY-Ae (YAe, Y-Ae)), Biotin, eBioscience™, Invitrogen™
Description
Description: The Y-Ae antibody reacts with a class II major histocompatibility complex (MHC) self Ea peptide (peptide 52-68) bound to I-Ab molecules. The Y-Ae antibody detects a determit expressed on a subset of class II I-Ab molecules in strains that also express class II I-Eb. This determit is expressed on peripheral B cells and on cells in the thymic medulla, but not thymic cortical epithelium. In addition to B cells, the Y-Ae determit is expressed at comparable levels on other antigen presenting cells, including macrophages and dendritic cells. The antibody does not react with invariant chain-associated class II MHC complexes. The reactivity of the Y-Ae antibody is MHC-restricted. The Y-Ae antibody was generated by immunization of [B10.MBR x B10.D2]F1 recipients with B10A(5R) LPS blasts. In this strain combination, there is incompatibility for I-Ab and I-Eb molecules. The antibody reacts with 10-15% of surface I-Ab molecules in strains that also express surface I-Eb molecules (e.g., strains B10.A(5R) and B10.A(3R)). The antibody does not react with I-A molecules in strains that have a non-functional I-E (a chain) gene (e.g., strains B6 and B10), nor with mutant I-A molecules (e.g., (B6.c-H-2bm-12 x A.TFR5)F1), nor with any other strain tested. The Y-Ae antibody recognizes an Ea peptide (Ea 52-68) bound to I-Ab molecules, but not to Ea 52-68 peptide bound to I-Ak or I-Abm-12 molecules.
Specifications
Specifications
| Antigen | Ea52-68 peptide bound to I-Ab |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | eBioY-Ae (YAe, Y-Ae), Y-Ae) |
| Concentration | 0.5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | H2-Ab1 |
| Gene Accession No. | P01921, P06342, P06343, P06344, P06345, P06346, P14483 |
| Gene Alias | Abeta; AI845868; H-2 class II histocompatibility antigen, A beta chain; H-2Ab; H2-Ab; H2-Ab1; H2Eb; H-2Eb; H2-iabeta; histocompatibility 2, class II antigen A, beta 1; Ia2; Ia-2; Ia4; Ia-4; IAb; I-Abeta; major histocompatibility complex class II beta chain; MGC163794; MGC163796; MHC class II antigen A beta; MHC class II H2-IA-beta-psi; response to metastatic cancers 1; Rmcs1 |
| Show More |
For Research Use Only.