missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E2F1 Recombinant Rabbit Monoclonal Antibody (23GB1505)

Catalog No. MA553965
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553965 20 μL
MA553964 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553965 Supplier Invitrogen™ Supplier No. MA553965

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

E2F1 Recombinant Rabbit Monoclonal Antibody (23GB1505)

E2F1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Specifications

Antigen E2F1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1505
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene E2F1
Gene Accession No. O09139, Q01094, Q61501
Gene Alias E2F transcription factor 1; E2F1; E2F-1; mKIAA4009; PBR3; PRB-binding protein E2F-1; RBAP1; RBAP-1; RBBP3; RBBP-3; RBP3; Retinoblastoma-associated protein 1; retinoblastoma-binding protein 3; Transcription factor E2F1
Gene Symbols E2F1
Host Species Rabbit
Immunogen A synthesized peptide derived from human E2F1 (355-400AA).
Purification Method Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13555, 1869, 399489
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less