missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F1 Recombinant Rabbit Monoclonal Antibody (23GB1505)

Description
E2F1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Spécifications
Spécifications
| Antigène | E2F1 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 23GB1505 |
| Concentration | 0.4 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gène | E2F1 |
| Numéro d’acquisition du gène | O09139, Q01094, Q61501 |
| Alias du gène | E2F transcription factor 1; E2F1; E2F-1; mKIAA4009; PBR3; PRB-binding protein E2F-1; RBAP1; RBAP-1; RBBP3; RBBP-3; RBP3; Retinoblastoma-associated protein 1; retinoblastoma-binding protein 3; Transcription factor E2F1 |
| Afficher plus de résultats |