missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ E1-ClipTip™ Electronic Adjustable Tip Spacing Multichannel Equalizer Pipettes
GREENER_CHOICE

Catalog No. 14387972BT
Change view
Click to view available options
Volume (Metric):
0.5 to 12.5 μL
1 to 30 μL
10 to 300 μL
15 to 1250 μL
2 to 125 μL
Number of Channels:
12
6
8
Compatible Tips:
ClipTip 1250
ClipTip 200
ClipTip 300
ClipTip 384 12.5
ClipTip 384 125
ClipTip 384 30
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Number of Channels Compatible Tips
14387972BT 0.5 to 12.5 μL 12 ClipTip 384 12.5
14387965BT 15 to 1250 μL 6 ClipTip 1250
14387966BT 2 to 125 μL 8 ClipTip 200
14387967BT 10 to 300 μL 8 ClipTip 300
14387968BT 15 to 1250 μL 8 ClipTip 1250
14387969BT 0.5 to 12.5 μL 8 ClipTip 384 12.5
14387970BT 1 to 30 μL 8 ClipTip 384 30
14387971BT 2 to 125 μL 8 ClipTip 384 125
14387973BT 1 to 30 μL 12 ClipTip 384 30
14387974BT 2 to 125 μL 12 ClipTip 384 125
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 14387972BT Supplier Thermo Scientific™ Supplier No. 4672020BT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ E1-ClipTip™ Equalizer Pipette can be conveniently used with any tube, rack, microplate or horizontal gel box due to the adjustable tip spacing

Perform sample transfers between virtually any tube, rack, microplate or horizontal gel box quickly and efficiently with the Thermo Scientific™ E1-ClipTip™ Equalizer Pipette. Adjustable tip spacing allows you to set the distance between tips by simply sliding the scale to expand or contract to the desired labware format.The NEW My Pipette™ Creator app enables programming of E1-ClipTip from the comfort of your PC

Innovative ClipTip interlocking tip interface technology*
  • Tip is locked and sealed in place until ejected — no dropped tips that waste valuable samples
  • Complete seal to secure the sample volume in each tip for enhanced accuracy and precision
  • Tips are attached and ejected with a light touch — eliminates the frustration of banging tips on the pipette

My Pipette Creator to revolutionize the way you pipette with a new era of technology
  • New web-based pipetting app powered by Thermo Fisher Cloud
  • Enables efficient, centralized programming and sharing of protocols between E1-ClipTip pipettes and colleagues
  • Offers pre-programmed protocols, to download and share, for many of your favorite reagent kits and pipetting protocols
  • Connect E1-ClipTip pipette to the app via a Bluetooth** or USB connection.
  • Accelerate collaboration and reproducibility, all from the comfort of your computer.

E1-ClipTip pipette to help you enjoy pipetting
  • E1-ClipTip is designed for optimum comfort and ease of use: the electronic tip ejection and index finger pipetting action let your thumb relax during pipetting.
  • Two operating options: Step-based Matrix or Presets with settings for your most common applications.
  • The Programs function allows you to store 30 your most common Matrix or Presets protocols with a specific name.
  • Connect your E1-ClipTip pipette to My Pipette Creator app conveniently via Bluetooth**
  • Five different user profiles offer customized settings for multiple users and protocols
  • MyPipette™ function to customize your main menu view by organizing the icons in your preferred order or creating shortcuts for your favorite programs and pipetting functions. You can also protect important settings from unintentional editing with a password.
  • Service and calibration trackers remind you when the pipette needs to be sent for service
  • Li-ion battery for long lasting performance and fast recharging. Recharging with a charging stand or a plug, you decide.
  • 2-year warranty with convenient web registration

E1-ClipTip™ Equalizer: transferring multiple samples between different labware formats
  • Adjustable tip spacing allows you to set the distance between pipette tips for different types of labware simply by sliding a scale to expand or contract the tip spacing to the desired format:
    • Microcentrifuge tubes to/from 24,-48,-96-and 384- well microplates
    • Test tubes to/from deepwell blocks
    • Microplates and tubes to agarose gels
  • Save up to 90% in time and repetitions compared to use of single channel manual pipettes***

E1-ClipTip pipette and ClipTip tips for every application
  • Choose your E1-ClipTip, volume range from 0.5 μL to 1250μL:
    • Single channels
    • Multichannels with 8, 12 or 16 channels in 96 and 384 formats
    • Adjustable tip spacing multichannels with 6, 8, 12 channels in 96 and 384 formats
  • Wide selection of ClipTip tips:
    • ClipTip 96-format tips are available in 20, 50, 200, 300, 1000 and 1250 μl
    • ClipTip 384-format tips are available in 12.5, 30 and 125 μl
    • Several packaging options: filtered, sterile, non-sterile, racked and reload

Compatible with Thermo Scientific ClipTip pipette tips.

* E1-ClipTip 12.5 and all 384 E1-ClipTip pipettes utilize snap and seal technology.
** Bluetooth models available in following countries/regions: Andorra, Australia, Austria, Belgium, Bulgaria, Canada, China, Croatia, Cyprus, Czech Republic, Denmark, Estonia, Finland, France, Germany, Greece, Hong Kong, Hungary, Iceland, India, Ireland, Italy, Japan, Korea (Rep. of), Latvia, Liechtenstein, Lithuania, Luxembourg, Malta, Monaco, Netherlands, New Zealand, Norway, Poland, Portugal, Romania, San Marino, Singapore, Slovakia, Slovenia, Spain, Sweden, Switzerland, Taiwan, United Kingdom, United States and Vatican. For other countries catalog numbers without 'BT' extension should be used.
*** Compared to a manual single channel pipette filling a 96-well microplate from microcentrifuge tubes. See Thermo Scientific Smart Note: Is there an easier and more efficient way to transfer liquids between various labware formats, than using traditional handheld pipettes? 2014.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications

Compatible Tips ClipTip 384 12.5
Description E1-ClipTip Equalizer
Adjustable Tip Spacing Yes
Color Pink
For Use With ClipTip 384 12.5
Model Bluetooth
Number of Channels 12
Product Line E1-ClipTip
Programmability Yes
Type Equalizer Pipette
Volume (Metric) 0.5 to 12.5 μL
Increments 0.01μL
Show More Show Less