missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human E-selectin (Soluble) ELISA Kit

Catalog No. BMS2052
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BMS2052 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BMS2052 Supplier Invitrogen™ Supplier No. BMS2052
Only null left

ELISA

Leukocytes adhere to the blood vessel endothelium during extravasation in postcapillary venules of lymph nodes. In addition, leukocyte adhesion occurs in the capillaries and small venules at any site in the body after onset of inflammation. This response is immediate and involves a cascade of adhesion receptors. At the endothelial surface members of the selectin and immunoglobulin superfamilies participate in this cascade. Selectins are C-type cell surface lectins that play a role in leukocyte adhesion to the blood vessel wall endothelium. E-selectin (CD62E) is an endothelial cell specific selectin that is expressed only after activation with proinflammatory cytokines. In vitro experiments have shown that IL-1, TNFalpha and bacterial wall components like lipopolysaccharides induce the transcription of E-selectin in a NFkB dependent signaling cascade. E-selectin has been associated with blood vessel endothelium in diverse inflammatory situations.

Specifications

Accession Number P16581
Assay Range 1.6 to 50 ng/mL
Assay Sensitivity 0.017 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 6401
Gene Symbol SELE
Interassay CV 3.3%
Intraassay CV 5.7
Kit Contents Pre-coated 96 well plate, Standard (2 vials), Sample diluent, Biotinylated Detection Antibody, Assay buffer Concentrate, Wash buffer concentrate, SAV-HRP, TMB, Stop solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 20 μL, Supernatant 20 μL
Storage Requirements 2°C to 8°C
Target E-selectin (CD62E)
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 30 mins
Show More Show Less