Learn More
Invitrogen™ E-Gel™ Double Comb Agarose Gels with SYBR™ Safe DNA Gel Stain

Description
Invitrogen E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain are designed for fast, convenient, and less-hazardous DNA sample electrophoresis. Each E-Gel Double Comb gel cassette contains all components and stain required for efficient gel separation and analysis–just load your samples and run. E-Gel Double Comb gels are ideal for analyzing PCR products, restriction digests, plasmid preparations, and genotyping samples.
Features of E-Gel Double Comb Agarose Gels include:
- Instantaneous setup—ready for use when you are and no gel preparation or additional buffers required
- Less-hazardous choice—contains SYBR Safe DNA stain, a less-hazardous alternative to ethidium bromide
- High performance—detect 3 ng of sample DNA with a separation time as short as 13–15 minutes
- In-process control—view DNA sample migration in real time
- Easy sample access—openable cassette design allows easy in-gel access for convenient DNA sample excision or transfer to membranes
- Better sample integrity—exploits blue-light excitation wavelength to minimize DNA damage, as opposed to UV
Less hazardous option
Choose SYBR Safe DNA gel stain to minimize exposure to hazardous and mutagenic options such as ethidium bromide and UV light. Precast E-Gel Double Comb gels with SYBR Safe stain, in combination with blue-light trans-illumination, offer better sample integrity and a less hazardous work environment.
Fast and convenient analysis
E-Gel agarose gels enable fast and high-performance analysis with a detection sensitivity of 3 ng of sample DNA per band and a separation time 2–3 fold faster than that of conventional pour-your-own gels. Each E-Gel Double Comb gel contains agarose, electrodes, SYBR Safe DNA stain, and a buffer-less ion exchange system packaged inside a dry disposable cassette. E-Gel technology does not require any gel or buffer preparation or gel staining steps. Just load your samples and run.
Live monitoring of DNA migration
E-Gel Double Comb gels include SYBR Safe DNA Gel Stain, which has an excitation wavelength in the blue-light spectrum. When used on the E-Gel Power Snap Electrophoresis Device with integrated blue-light trans-illuminator, the gels enable less hazardous real-time monitoring of DNA migration without risk to the eyes or risk of damaging the sample, as opposed to UV visualization.
Easy access to the gel
E-Gel Double Comb cassettes are designed for convenient opening with a Gel Knife (Cat. No. EI9010), so that the gel inside can be accessed readily for excision of specific bands or transferred to a membrane for southern blot analysis.
Running and imaging devices
E-Gel Double Comb gels require the E-Gel Power Snap Electrophoresis System for gel running and viewing. The system consists of an electrophoresis device and a camera for fast and convenient E-Gel agarose gel separation and analysis.
Available in a variety of gel formats
E-Gel agarose gels are available in variety of gel percentage, stain, and well formats. The 22-well E-Gel Double Comb gels with SYBR Safe stain are available in 1% and 2% gel percentages.
Specifications
Specifications
| Compatible Ladders | E-Gel™ 1 Kb Plus Express Ladder |
| Content And Storage | • Each box contains 10 gels Store strictly at room temperature. Do not freeze. Avoid cold spots near freezers or other uncontrolled environments. |
| Gel Percentage | 1% |
| Gel Type | E-Gel |
| Label or Dye | SYBR Safe |
| Separation Range | 100 to 5000 bp |
| Wells | 22-well |
| Quantity | 5 x 10 Gels |
| Well Design | Double Comb |
| Product Type | Agarose Gel |
For Research Use Only. Not for use in diagnostic procedures.