missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E. coli (strain K12/DH10B) NfuA Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144415
Description
E. coli (strain K12/DH10B) NfuA Polyclonal Antibody for Western Blot, ELISA
Involved in iron-sulfur cluster biogenesis. Binds a 4Fe-4S cluster, can transfer this cluster to apoproteins, and thereby intervenes in the maturation of FeS proteins. Could also act as a scaffoldchaperone for damaged FeS proteins.Chemical Identifiers
| E. coli (strain K12/DH10B) NfuA | |
| -20°C or -80°C if preferred | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Bacteria | |
| nfuA | |
| Fe/S biogenesis protein NfuA nfuA ECDH10B_3589 | |
| nfuA | |
| Primary | |
| Protein G |
| 2.88 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| B1X760 | |
| 58390684, 82263228 | |
| Recombinant Escherichia coli (strain K12/DH10B) NFUA protein (31-128aa). | |
| Antibody |
Specifications
| E. coli (strain K12/DH10B) NfuA | |
| Polyclonal | |
| Unconjugated | |
| nfuA | |
| Fe/S biogenesis protein NfuA nfuA ECDH10B_3589 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 58390684, 82263228 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2.88 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| B1X760 | |
| nfuA | |
| Recombinant Escherichia coli (strain K12/DH10B) NFUA protein (31-128aa). | |
| 100 μL | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |