missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Cadherin Monoclonal Antibody (4A2C7)
GREENER_CHOICE

Catalog No. 180223
Encompass
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
180223 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 180223 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' 180223

Mouse Monoclonal Antibody

This monoclonal antibody reacts with human epithelial cadherin (E-cadherin, uvomorulin, Cell-CAM 120/80), a 120 kDa transmembrane glycoprotein. Reduction and/or loss of E-cadherin expression in carcinomas correlates positively with tumor cell invasion and metastasis. Altered expression of E-cadherin is common in the progression of cancers including breast carcinoma, lung carcinoma, gastric tumors, hepatocellular carcinoma, and prostatic tumors. It has recently been reported that E-cadherin is a useful marker for differentiating lobular lesions (negative) from ductal invasive lesions (positive) in breast cancer. Positive tissue: Non-neural epithelia. Reacts strongly with human E-cadherin and weakly with human P-cadherin. Expected Staining Pattern: Mainly cellular membrane, some cytoplasmic.

E-Cadherin (epithelial cadherin) is a classical cadherin from the cadherin (alcium dependent adhesion protein) superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid and ovarian cancer. Loss of function is thought to contribute to progression in cancer by increasing proliferation, invasion, and/or metastasis. The ectodomain of this protein mediates bacterial adhesion to mammalian cells and the cytoplasmic domain is required for internalization. Identified transcript variants arise from mutation at consensus splice sites. E-cadherin plays a central role in the growth and development of cells by controlling tissue architecture, and maintenance of tissue integrity. In humans, E-cadherin is encoded by the CDH1 gene present on chromosome 16. Studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.

Specifications

Antigen E-Cadherin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4A2C7
Conjugate Unconjugated
Dilution Concentrate Antibody
Formulation PBS with 1% BSA and 0.1% sodium azide
Gene CDH1
Gene Accession No. P12830
Gene Alias AA960649; ARC-1; cadherin 1; cadherin 1, E-cadherin (epithelial); cadherin 1, epithelial; cadherin 1, type 1; cadherin 1, type 1, E-cadherin (epithelial); cadherin e; cadherin-1; cadherin-E; calcium-dependent adhesion protein, epithelial; CAM 120/80; CD324; CDH1; CDHE; cell adhesion molecule; cell-CAM 120/80; ECAD; E-cad; E-Cad/CTF1; E-Cad/CTF2; E-Cad/CTF3; Ecadherin; E-cadherin; E-cadherin 1; E-cadherin precursor; Epithelial cadherin; hab; half baked; LCAM; L-CAM; Um; UVO; Uvomorulin
Gene Symbols CDH1
Host Species Mouse
Immunogen Recombinant protein corresponding to the cytoplasmic domain of human E-cadherin.
Positive Control Non-neural epithelia
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 999
Packaging 1 Each
Test Specificity E-Cadherin
Includes 0.1% Sodium azide
Target Species Human
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Host abrv Ms
Reactivity Human
Shipping Condition Wet Ice
Reactivity abrv Hu
Target Gene E-Cadherin
Validated Application abrv IHC (FFPE)
Show More Show Less