missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals Inc E-64, ∽99%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215284601

 View more versions of this product

Catalog No. ICN15284601



Description

Description

Specifications

Specifications

E-64
White
C15H27N5O5
1 mg
LTLYEAJONXGNFG-HBNTYKKESA-N
(2R,3R)-3-{[(1S)-1-({4-[(diaminomethylidene)amino]butyl}carbamoyl)-3-methylbutyl]carbamoyl}oxirane-2-carboxylic acid
4641099
Powder
66701-25-5
∽99%
MFCD00080261,MFCD00080261
d09fgq, l-3-trans-carboxy-oxiran-2-carbonyl-leu-agmatin, trans-epoxysuccinyl-leucylamido-4-guanidino butane, 3-1-4-carbamimidamidobutyl carbamoyl-3-methylbutyl carbamoyl oxirane-2-carboxylic acid, 3-1-4-diaminomethylideneamino butylamino-4-methyl-1-oxopentan-2-yl carbamoyl oxirane-2-carboxylic acid
CC(C)C[C@H](NC(=O)[C@@H]1O[C@H]1C(O)=O)C(=O)NCCCCN=C(N)N
357.41
357.4

Chemical Identifiers

66701-25-5
357.41
LTLYEAJONXGNFG-HBNTYKKESA-N
4641099
CC(C)C[C@H](NC(=O)[C@@H]1O[C@H]1C(O)=O)C(=O)NCCCCN=C(N)N
C15H27N5O5
MFCD00080261,MFCD00080261
d09fgq, l-3-trans-carboxy-oxiran-2-carbonyl-leu-agmatin, trans-epoxysuccinyl-leucylamido-4-guanidino butane, 3-1-4-carbamimidamidobutyl carbamoyl-3-methylbutyl carbamoyl oxirane-2-carboxylic acid, 3-1-4-diaminomethylideneamino butylamino-4-methyl-1-oxopentan-2-yl carbamoyl oxirane-2-carboxylic acid
(2R,3R)-3-{[(1S)-1-({4-[(diaminomethylidene)amino]butyl}carbamoyl)-3-methylbutyl]carbamoyl}oxirane-2-carboxylic acid
SDS
Documents

Documents

Promotions

Promotions