missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(E)-4-Methoxycinnamaldehyde 97.0+%, TCI America™
Supplier: TCI America M101225G
Specifications
| (E)-4-Methoxycinnamaldehyde | |
| 58°C | |
| 176°C | |
| MFCD00017343 | |
| 4-methoxycinnamaldehyde, 3-4-methoxyphenyl acrylaldehyde, p-methoxycinnamaldehyde, e-3-4-methoxyphenyl acrylaldehyde, 3-4-methoxyphenyl-2-propenal, trans-4-methoxycinnamaldehyde, p-methoxy cinnamaldehyde, p-methoxycinnamic aldehyde, trans-p-methoxycinnamaldehyde, cinnamaldehyde, p-methoxy | |
| COC1=CC=C(\C=C\C=O)C=C1 | |
| 162.19 | |
| CHEBI:27952 | |
| ≥97.0% (GC) |
| 24680-50-0 | |
| Yellow | |
| C10H10O2 | |
| 25 g | |
| AXCXHFKZHDEKTP-NSCUHMNNSA-N | |
| (2E)-3-(4-methoxyphenyl)prop-2-enal | |
| 641294 | |
| 162.19 | |
| Crystalline Powder |
Chemical Identifiers
| 24680-50-0 | |
| 162.19 | |
| AXCXHFKZHDEKTP-NSCUHMNNSA-N | |
| 641294 | |
| (2E)-3-(4-methoxyphenyl)prop-2-enal |
| C10H10O2 | |
| MFCD00017343 | |
| 4-methoxycinnamaldehyde, 3-4-methoxyphenyl acrylaldehyde, p-methoxycinnamaldehyde, e-3-4-methoxyphenyl acrylaldehyde, 3-4-methoxyphenyl-2-propenal, trans-4-methoxycinnamaldehyde, p-methoxy cinnamaldehyde, p-methoxycinnamic aldehyde, trans-p-methoxycinnamaldehyde, cinnamaldehyde, p-methoxy | |
| CHEBI:27952 | |
| COC1=CC=C(\C=C\C=O)C=C1 |
Safety and Handling
TSCA : Yes