missing translation for 'onlineSavingsMsg'
Learn More

(E)-4-Methoxycinnamaldehyde 97.0+%, TCI America™

Catalog No. p-7132496
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10125G 5 g
M101225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10125G Supplier TCI America Supplier No. M10125G
Only null left

Chemical Identifiers

CAS 24680-50-0
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00017343
InChI Key AXCXHFKZHDEKTP-NSCUHMNNSA-N
Synonym 4-methoxycinnamaldehyde, 3-4-methoxyphenyl acrylaldehyde, p-methoxycinnamaldehyde, e-3-4-methoxyphenyl acrylaldehyde, 3-4-methoxyphenyl-2-propenal, trans-4-methoxycinnamaldehyde, p-methoxy cinnamaldehyde, p-methoxycinnamic aldehyde, trans-p-methoxycinnamaldehyde, cinnamaldehyde, p-methoxy
PubChem CID 641294
ChEBI CHEBI:27952
IUPAC Name (2E)-3-(4-methoxyphenyl)prop-2-enal
SMILES COC1=CC=C(\C=C\C=O)C=C1

Specifications

Melting Point 58°C
Color Yellow
Boiling Point 176°C
Quantity 5 g
Formula Weight 162.19
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (E)-4-Methoxycinnamaldehyde