missing translation for 'onlineSavingsMsg'
Learn More

(E)-4-Acide aminocinnamique 98,0+%, TCI America™

Numéro de catalogue. p-7117007
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A06915G 5 g
A069125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. A06915G Fournisseur TCI America Code fournisseur A06915G

Identifiants chimiques

CAS 17570-30-8
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00017117
InChI Key JOLPMPPNHIACPD-ZZXKWVIFSA-N
Synonym (E)-3-(4-Aminophenyl)acrylic Acid
PubChem CID 1549514
IUPAC Name (E)-3-(4-aminophényl)prop-2-acide énoïque
SMILES C1=CC(=CC=C1C=CC(=O)O)N

Spécifications

Melting Point 170°C
Quantity 5 g
Formula Weight 163.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Poudre cristalline
Chemical Name or Material (E)-4-Aminocinnamic Acid
RTECSNumber UD3000000
TSCA Yes