accessibility menu, dialog, popup

UserName

OHONH2(E/Z)

CAS RN 17570-30-8

IUPAC Name: 3-(4-aminophenyl)prop-2-enoic acid
Synonyms: 3-(4-aminophenyl)prop-2-enoic acid
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: JOLPMPPNHIACPD-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=CC(O)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results