missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(E)-4-Aminocinnamic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 170°C |
| Quantity | 25 g |
| Formula Weight | 163.18 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (E)-4-Aminocinnamic Acid |
Safety and Handling
| RTECSNumber | UD3000000 |
| TSCA | Yes |