missing translation for 'onlineSavingsMsg'
Learn More

(E)-4-Aminocinnamic Acid 98.0+%, TCI America™

Catalog No. A06915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A06915G 5 g
A069125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A06915G Supplier TCI America Supplier No. A06915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17570-30-8
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00017117
InChI Key JOLPMPPNHIACPD-ZZXKWVIFSA-N
Synonym (E)-3-(4-Aminophenyl)acrylic Acid
PubChem CID 1549514
IUPAC Name (E)-3-(4-aminophenyl)prop-2-enoic acid
SMILES C1=CC(=CC=C1C=CC(=O)O)N

Specifications

Melting Point 170°C
Quantity 5 g
Formula Weight 163.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (E)-4-Aminocinnamic Acid
RTECSNumber UD3000000
TSCA Yes