missing translation for 'onlineSavingsMsg'
Learn More

(E)-4-(4-Hydroxy-3-methoxyphenyl)-3-buten-2-one 98.0+%, TCI America™

Catalog No. p-7130851
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09965G 5 g
H099625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09965G Supplier TCI America Supplier No. H09965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22214-42-2
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.21
MDL Number MFCD00012210
InChI Key AFWKBSMFXWNGRE-UHFFFAOYSA-N
Synonym Dehydrozingerone, Vanillylideneacetone
PubChem CID 5354238
ChEBI CHEBI:81361
IUPAC Name 4-(4-hydroxy-3-methoxyphenyl)but-3-en-2-one
SMILES COC1=C(O)C=CC(C=CC(C)=O)=C1

Specifications

Melting Point 129°C
Quantity 5 g
Formula Weight 192.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (E)-4-(4-Hydroxy-3-methoxyphenyl)-3-buten-2-one
RTECSNumber EM9960000
TSCA Yes