missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(E)-4-(4-Hydroxy-3-methoxyphenyl)-3-buten-2-one 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 129°C |
| Quantity | 25 g |
| Formula Weight | 192.21 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (E)-4-(4-Hydroxy-3-methoxyphenyl)-3-buten-2-one |
Safety and Handling
| RTECSNumber | EM9960000 |
| TSCA | Yes |