missing translation for 'onlineSavingsMsg'
Learn More

(E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein 98.0+%, TCI America™

Catalog No. F08195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08195G 5 g
F081925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08195G Supplier TCI America Supplier No. F08195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 93957-50-7
Molecular Formula C20H18FNO
Molecular Weight (g/mol) 307.368
MDL Number MFCD03840863
InChI Key DVWHSTKQJBIYCK-VMPITWQZSA-N
Synonym (E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]propenal
PubChem CID 9926593
IUPAC Name (E)-3-[3-(4-fluorophenyl)-1-propan-2-ylindol-2-yl]prop-2-enal
SMILES CC(C)N1C2=CC=CC=C2C(=C1C=CC=O)C3=CC=C(C=C3)F

Specifications

Melting Point 131°C
Color Yellow
Quantity 5 g
UN Number 3077
Formula Weight 307.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein