missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) trifluoromethanesulfonate, 98%

CAS: 139177-62-1 | C3DyF9O9S3 | 609.69 g/mol

Supplier:  Thermo Scientific Chemicals L2025214

 View more versions of this product

Catalog No. AAL2025214


Only null left
Explore available promotions

Description

Description

Dysprosium(III) trifluoromethanesulfonate is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes. This is an effective catalyst for electrophilic substitution reactions of indoles with imines. Also used as catalyst for Aza-Piancatelli rearrangement, Friedel-Crafts alkylation, ring-opening polymerization reactions, microwave-assisted Kabachnik-Fields condensation, cycloaddition reactions (Lewis-acid catalyst), fries rearrangement and enantioselective glyoxalate-ene reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dysprosium(III) trifluoromethanesulfonate is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes. This is an effective catalyst for electrophilic substitution reactions of indoles with imines. Also used as catalyst for Aza-Piancatelli rearrangement, Friedel-Crafts alkylation, ring-opening polymerization reactions, microwave-assisted Kabachnik-Fields condensation, cycloaddition reactions (Lewis-acid catalyst), fries rearrangement and enantioselective glyoxalate-ene reactions.

Solubility
Soluble in water.

Notes
Hygroscopic. store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Dysprosium(III) trifluoromethanesulfonate
C3DyF9O9S3
MFCD00209583
Hygroscopic
Soluble in water.
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Dy+3]
609.69
609.69
139177-62-1
Dy(SO3CF3)3
25 g
dysprosium iii trifluoromethanesulfonate, tris triflato dysprosium, dy otf 3, dysprosium iii triflate, dysprosium triflate, dysprosium trifluoromethanesulfonate, acmc-20al36, dysprosium 3+ tritriflate, dysprosium tris trifluoromethanesulfonate, dysprosium 3+ ; trifluoromethanesulfonate
XSVCYDUEICANRJ-UHFFFAOYSA-K
dysprosium(3+);trifluoromethanesulfonate
9938788
98%

Chemical Identifiers

139177-62-1
609.69
XSVCYDUEICANRJ-UHFFFAOYSA-K
9938788
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Dy+3]
C3DyF9O9S3
MFCD00209583
dysprosium iii trifluoromethanesulfonate, tris triflato dysprosium, dy otf 3, dysprosium iii triflate, dysprosium triflate, dysprosium trifluoromethanesulfonate, acmc-20al36, dysprosium 3+ tritriflate, dysprosium tris trifluoromethanesulfonate, dysprosium 3+ ; trifluoromethanesulfonate
dysprosium(3+);trifluoromethanesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335-H500

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only