CAS RN 139177-62-1
IUPAC Name:
dysprosium(3+) tritrifluoromethanesulfonate
Synonyms:
dysprosium(3+) tritriflate
Molecular Weight (g/mol):
609.69
Molecular Formula:
C3DyF9O9S3
InChi Key:
XSVCYDUEICANRJ-UHFFFAOYSA-K
SMILES:
[Dy+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
Dysprosium(III) trifluoromethanesulfonate, 98%
Dysprosium(III) trifluoromethanesulfonate, 98%
CAS: 139177-62-1 Molecular Formula: C3DyF9O9S3 Molecular Weight (g/mol): 609.69 MDL Number: MFCD00209583 InChI Key: XSVCYDUEICANRJ-UHFFFAOYSA-K Synonym: dysprosium iii trifluoromethanesulfonate,tris triflato dysprosium,dy otf 3,dysprosium iii triflate,dysprosium triflate,dysprosium trifluoromethanesulfonate,acmc-20al36,dysprosium 3+ tritriflate,dysprosium tris trifluoromethanesulfonate,dysprosium 3+ ; trifluoromethanesulfonate PubChem CID: 9938788 IUPAC Name: dysprosium(3+);trifluoromethanesulfonate SMILES: C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Dy+3]