missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) trifluoromethanesulfonate, 98%

Catalog No. AAL2025206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2025206 5 g
AAL2025214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2025206 Supplier Thermo Scientific Chemicals Supplier No. L2025206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 139177-62-1 | C3DyF9O9S3 | 609.69 g/mol

Dysprosium(III) trifluoromethanesulfonate is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes. This is an effective catalyst for electrophilic substitution reactions of indoles with imines. Also used as catalyst for Aza-Piancatelli rearrangement, Friedel-Crafts alkylation, ring-opening polymerization reactions, microwave-assisted Kabachnik-Fields condensation, cycloaddition reactions (Lewis-acid catalyst), fries rearrangement and enantioselective glyoxalate-ene reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dysprosium(III) trifluoromethanesulfonate is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes. This is an effective catalyst for electrophilic substitution reactions of indoles with imines. Also used as catalyst for Aza-Piancatelli rearrangement, Friedel-Crafts alkylation, ring-opening polymerization reactions, microwave-assisted Kabachnik-Fields condensation, cycloaddition reactions (Lewis-acid catalyst), fries rearrangement and enantioselective glyoxalate-ene reactions.

Solubility
Soluble in water.

Notes
Hygroscopic. store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 139177-62-1
Molecular Formula C3DyF9O9S3
Molecular Weight (g/mol) 609.69
MDL Number MFCD00209583
InChI Key XSVCYDUEICANRJ-UHFFFAOYSA-K
Synonym dysprosium iii trifluoromethanesulfonate, tris triflato dysprosium, dy otf 3, dysprosium iii triflate, dysprosium triflate, dysprosium trifluoromethanesulfonate, acmc-20al36, dysprosium 3+ tritriflate, dysprosium tris trifluoromethanesulfonate, dysprosium 3+ ; trifluoromethanesulfonate
PubChem CID 9938788
IUPAC Name dysprosium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Dy+3]

Specifications

Linear Formula Dy(SO3CF3)3
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 609.69
Percent Purity 98%
Chemical Name or Material Dysprosium(III) trifluoromethanesulfonate
Hazard Category H314-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only