missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific™

Catalog No. AA4140918
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4140918 50 g
AA4140909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4140918 Supplier Thermo Scientific Chemicals Supplier No. 4140918

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For vacuum deposition

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 13569-80-7
MDL Number MFCD00016071

Specifications

Boiling Point 2200°C
Melting Point 1360°C
Color White
Physical Form Pieces
Quantity 50 g
Solubility Information Soluble in strong mineral acids. Insoluble in water.
Formula Weight 219.5
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 7.5 g/mL
Assay (REO)
Chemical Name or Material Dysprosium(III) fluoride, Anhydrous
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-992-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen