missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dysprosium ICP Standard, 1000 ppm Dy in 3% HNO3, Ricca Chemical
Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| Concentration or Composition Notes | 1,000ppm Dy |
| CAS Min % | 93.0, 4.63, 0.11 |
| CAS Max % | 97.0, 4.81, 0.11 |
| Equivalence | 1 mL = 1 mg Dy |
| Chemical Name or Material | Dysprosium ICP Standard |