missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dysferlin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554541
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554541 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554541 Supplier Invitrogen™ Supplier No. PA554541
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: CHYYYLPWGN VKPVVVLSSY WEDISHRIET QNQLLGIADR LEAGLEQVHL ALKAQCSTED VDSLVAQLTD ELIAGCSQPL GDIHETPSAT HLDQYLYQLR THHLSQITEA ALALKLGHSE LPAALEQAED WLLRLRALA Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

Dysferlin is the protein product of the 2p13 gene that is defective in patients with Limb-Girdle Muscular Dystrophy type 2B (LGMD2B) and Miyoshi Myopathy (MM). Dysferlin is normally localized to the muscle plasma membrane. In patients with LGMD2B and MM, immunoreactivity to dysferlin is severely reduced or lost, depending on the type of mutation. This antibody is used for the characterization of LGMD2B and MM.

Specifications

Antigen Dysferlin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DYSF
Gene Accession No. O75923
Gene Alias 2310004N10Rik; AI604795; D6Pas3; Dysf; Dysferlin; dysferlin variant a; dysferlin_a; dystrophy-associated fer-1-like 1; dystrophy-associated fer-1-like protein; FER1L1; fer-1-like family member 1; fer-1-like protein 1; FLJ00175; FLJ90168; LGMD2B; limb girdle muscular dystrophy 2B (autosomal recessive); MMD1
Gene Symbols DYSF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Dysferlin. Recombinant protein control fragment (Product #RP-90255).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8291
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less