missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DYRK4, GST Tag Recombinant Protein

Catalog No. PV3871
Encompass
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3871 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV3871 Supplier Invitrogen™ Supplier No. PV3871

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

Dual-specificity kinases, such as DYRK4, play key roles in cell proliferation, survival, and development.

Specifications

Accession Number Q9NR20
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.5mM EDTA, 50% glycerol, 0.02% Triton X-100, 2mM DTT, 150mM NaCl and no preservative; pH 7.5
Gene ID (Entrez) 8798
Molecular Weight (g/mol) 88.8 kDa
Name Human DYRK4, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AW049118; dual specificity tyrosine phosphorylation regulated kinase 4; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 4; dual specificity tyrosine-phosphorylation-regulated kinase 4; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 4; dyrk4; Dyrk4a; Dyrk4b; RAD51 associated protein 1; Rad51ap1
Common Name DYRK4
Gene Symbol Dyrk4
Kinase Family DYRK Kinase Family, DYRKs
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Assay Entry Biochemical terbium phospho-specific activity, Biochemical phospho-specific activity (no antibody)
Kinase Group Ser⁄Thr Kinases: CMCG Group
Shipping Condition Dry Ice
System Compatibility LanthaScreen™ Antibody, LanthaScreen™ Substrate, Zin-LYTE™
Target Entry DYRK4
Target Gene dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 4
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Full Length, Recombinant
Form Liquid
Protein Subtype Serine⁄Threonine Kinases
Research Category Signal Transduction
Purity or Quality Grade 55% by SDS-PAGE
Show More Show Less