missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Dynamis™ Medium

Catalog No. p-9726007
Change view
Click to view available options
Quantity:
1000 mL
10 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2661501 1000 mL
A2661502 10 L
A2661503 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A2661501 Supplier Gibco™ Supplier No. A2661501
Only null left

Gibco Dynamis liquid medium is specifically designed to offer the highest batch and fed-batch culture performance and yield with recombinant CHO cells in a chemically defined environment.

The chemically defined, protein-free, animal origin component-free formulation provides the power to achieve high titers, start process development faster and streamLine or simplify transfer to manufacturing scale.

Power to achieve higher titers.

Dynamis Medium is fortified to help maximize recombinant CHO cell growth and protein expression in suspension batch culture, offering 170% greater production than select competitive media with simple glucose supplementation by day 14. Cultures grown in Dynamis Medium extend to even higher titers demonstrating good cell viability out to day 21. This medium can also provide up to 60% greater fed-batch growth and production over glucose-only fed-batch culture for selected cell lines and applications when used with EfficientFeed C+ Supplement.

Power to start process development faster.

As a fortified medium, Dynamis provides the power to get to a high cell density over 10 million cells per mL faster than competitive products and sustains that cell density longer by just maintaining glucose levels. The viable cell growth achieved make it a good choice for scale-up if you have started with CD FortiCHO medium. When paired with EfficientFeed C+ Supplement, it provides a fast and efficient way to develop a process.

When to Use Dynamis Medium

  • The cell line you are using is a transfected CHO K1, GS CHO or CHO-S cell line
  • Maximum batch culture cell densities and protein titers are needed
  • You have time to adapt cells into a new medium in an effort to maximize titers
  • Cell health at harvest is a priority for downstream processing

Specifications

Cell Line CHO DG44, CHO-S, cGMP Banked, CHO-S, CHO-K1, GS-CHO, HEK293
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Liquid
Product Type AGT Medium
Product Line Dynamis
Quantity 1000 mL
Shipping Condition Room Temperature
Format Bottle(s)
Content And Storage Liquid format medium. Store in the dark at 2–8°C.
Culture Type Mammalian Cell Culture

For Research Use Only. Not for use in diagnostic procedures.