missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynamin 3 Recombinant Rabbit Monoclonal Antibody (23GB5175)

Catalog No. MA552809
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552809 20 μL
MA552808 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552809 Supplier Invitrogen™ Supplier No. MA552809
Only null left

Rabbit Recombinant Monoclonal Antibody

Dynamin 3 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

The dynamins are a family of 100 kDa GTPases transcribed from at least three separate genes. At least four mRNA splice variants for each dynamin have been described. Dynamins contain several conserved regions including the conserved, amino-terminal GTPase domain, a centrally located membrane-binding plekstrin homology domain (PHD), and a coiled-coil region located in front of a proline-rich domain (PRD). The PRD is thought to mediate interactions between dynamin and numerous other cellular proteins. Dynamin I is expressed exclusively in neurons, dynamin II is ubiquitously expressed, and dynamin III is thought to be restricted to expression in the brain, testis, heart, and lung. The dynamins participate in the cellular process of clathrin-mediated and fluid-phase endocytosis.

Specifications

Antigen Dynamin 3
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5175
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Dnm3
Gene Accession No. Q08877, Q8BZ98, Q9UQ16
Gene Alias 9630020E24Rik; AW061159; B230343F03Rik; C530045C17; DNM3; Dyn3; Dyna III; dynamin 3; dynamin family member; Dynamin III; dynamin, testicular; dynamin-2; Dynamin-3; KIAA0820; mKIAA0820; T-dynamin; testicular dynamin
Gene Symbols Dnm3
Host Species Rabbit
Immunogen A synthesized peptide derived from human Dynamin 3 (750-800AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103967, 171574, 26052
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less