missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynamic Diagnostics Isolation Station
Provides easy access to Personal Protection Equipment (PPE) in one area
$1093.57 - $1093.57
Specifications
| For Use With (Application) | Provides easy access to personal protection equipment in one area |
|---|---|
| Includes | Hardware |
| Color | White |
Includes: Hardware
| Catalog Number | Mfr. No. | Material | Product Type | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | Product Type | Price | Quantity | ||||
22-899-013
|
Dynamic Diagnostics
300019 |
Powder-coated Steel | PCW Isolation Station |
Each for $1,093.57
|
|
||||
Description
- Flat design allows for room where space is limited
- Optional Over-the-Door Hangers (sold separately) allow these units to be placed instantly when precautions are required
- Provides room for three glove boxes, folded gowns and facemasks
- 22G steel with rust-resistant white powder-coated finish
- Feature key holes for wall mounting for easy placement and removal of supplies
Specifications
| Provides easy access to personal protection equipment in one area | |
| Hardware | |
| White |