missing translation for 'onlineSavingsMsg'
Learn More

Dynamic Diagnostics 20-Place Aluminum Slide Tray DFS Item

Aluminum slide tray

Supplier:  Dynamic Diagnostics 300030

Catalog No. 22-899-108


Only null left

Description

Description

Aluminum slide tray

  • Made of aluminum
  • Tray dimensions: 11.25 x 7.25 x 0.25 in.
  • Holds 20 each of 25 x 75 x 1mm slides
Specifications

Specifications

Aluminum
11.25 in.
Rectangular
Slide Tray
0.25 in.
7.25 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions